missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tellurium(IV) oxide, tech., Thermo Scientific™

$208.49 - $777.10

Chemical Identifiers

CAS 7446-07-3
Molecular Formula O2Te
Molecular Weight (g/mol) 159.60
MDL Number MFCD00011263
InChI Key LAJZODKXOMJMPK-UHFFFAOYSA-N
Synonym 6-chlorobutyric acid, acmc-1bglq, oxotellane oxide, telluride oxide, tellurite teo2, tellurium iv oxide, tellurium oxide teo2, teo2, unii-397e9rke83
PubChem CID 62638
IUPAC Name tellurium dioxide
SMILES O=[Te]=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1060818
View Documents Promotion Details
Thermo Fisher Scientific
1060818
50g
Each for $208.49
 
AA1060830
View Documents Promotion Details
Thermo Fisher Scientific
1060830
250g
Each for $777.10
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7446-07-3
159.60
LAJZODKXOMJMPK-UHFFFAOYSA-N
62638
O=[Te]=O
O2Te
MFCD00011263
6-chlorobutyric acid, acmc-1bglq, oxotellane oxide, telluride oxide, tellurite teo2, tellurium iv oxide, tellurium oxide teo2, teo2, unii-397e9rke83
tellurium dioxide

Specifications

7446-07-3
MFCD00011263
Practically insoluble in water. Soluble in NaOH solutions and HCl
O=[Te]=O
159.60
159.6
Technical
733°C
Tellurium(IV) Oxide
O2Te
6-chlorobutyric acid, acmc-1bglq, oxotellane oxide, telluride oxide, tellurite teo2, tellurium iv oxide, tellurium oxide teo2, teo2, unii-397e9rke83
LAJZODKXOMJMPK-UHFFFAOYSA-N
tellurium dioxide
62638
Powder
5.8
50g
Safety and Handling

Safety and Handling

P201-P202-P261-P271-P272-P280g-P281-P302+P352-P304+P340-P308+P313-P333+P313-P363-P501c

H317-H332-H360

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 231-193-1

RTECSNumber : WY2675000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Product Certifications

RUO – Research Use Only