missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tellurium(IV) Oxide, 99.9995% (Metal Basis), Alfa Aesar™, Puratronic™

Manufacturer:  Thermo Fisher Scientific 1088236

 View more versions of this product

Catalog No. AA1088236



Specifications

Specifications

Tellurium(IV) Oxide
99.9995%, metal basis
MFCD00011263
LAJZODKXOMJMPK-UHFFFAOYSA-N
tellurium dioxide
62638
Powder
500g
7446-07-3
O2Te
6-chlorobutyric acid
O=[Te]=O
159.60
159.6
5.8

Chemical Identifiers

7446-07-3
159.60
LAJZODKXOMJMPK-UHFFFAOYSA-N
62638
O=[Te]=O
O2Te
MFCD00011263
6-chlorobutyric acid
tellurium dioxide
Documents