missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tellurium(IV) Oxide, 99.9995% (Metal Basis), Alfa Aesar™, Puratronic™

Manufacturer:  Thermo Fisher Scientific 1088214

 View more versions of this product

Catalog No. AA1088214



Specifications

Specifications

Tellurium(IV) Oxide
99.9995%, metal basis
MFCD00011263
Soluble in strong acids and alkali. Insoluble in water.
O=[Te]=O
159.60
159.6
5.8
25g
7446-07-3
O2Te
6-chlorobutyric acid, acmc-1bglq, oxotellane oxide, telluride oxide, tellurite teo2, tellurium iv oxide, tellurium oxide teo2, teo2, unii-397e9rke83
LAJZODKXOMJMPK-UHFFFAOYSA-N
tellurium dioxide
62638
Powder
733°C

Chemical Identifiers

7446-07-3
159.60
LAJZODKXOMJMPK-UHFFFAOYSA-N
62638
O=[Te]=O
O2Te
MFCD00011263
6-chlorobutyric acid, acmc-1bglq, oxotellane oxide, telluride oxide, tellurite teo2, tellurium iv oxide, tellurium oxide teo2, teo2, unii-397e9rke83
tellurium dioxide
Safety and Handling

Safety and Handling

P201-P202-P261-P271-P272-P280g-P281-P302+P352-P304+P340-P308+P313-P333+P313-P363-P501c

H317-H332-H360

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 231-193-1

RTECSNumber : WY2675000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents