Please login to your online account to display your discounted pricing

Tellurium(IV) oxide, 99.995% (metals basis), Alfa Aesar™

$974.88 - $974.88

Chemical Identifiers

Chemical Name or Material Tellurium(IV) oxide
CAS 7446-07-3
Name Note (metals basis)
Molecular Formula TeO2
Formula Weight 159.6
MDL Number MFCD00011263
Merck Index 14,9123
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1182130

  SDS
alfa aesar
1182130
Each for $974.88
Description & Specifications

Specifications

Formula Weight 159.6
Quantity 250g
Chemical Name or Material Tellurium(IV) oxide
TSCA TSCA
Odor Odorless
Name Note (metals basis)
Physical Form Powder
Assay 99.995%
Molecular Formula TeO2
MDL Number MFCD00011263
CAS 7446-07-3
Merck Index 14,9123
Melting Point 733°C
Density 5.8
RTECS Number WY2675000