Please login to your online account to display your discounted pricing

Alfa Aesar™ Tellurium(IV) isopropoxide, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Tellurium(IV) isopropoxide
CAS 1795-64-8
Formula Weight 363.73
MDL Number MFCD00145354
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3656502

  SDS
alfa aesar™
3656502
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 363.73
UN Number UN1993
Health Hazard 1 Warning
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
CAS 1795-64-8
Odor Alcohol-like
Health Hazard 2 GHS H Statement
H226-H302-H312-H332-H315-H319
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Sensitivity Moisture & Light Sensitive
Assay Percent Notes (metals basis)
Chemical Name or Material Tellurium(IV) isopropoxide
Boiling Point 76°C (0.5mmHg)
Assay 99.9%
MDL Number MFCD00145354
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 0.5g
Physical Form Liquid
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H302-H312-H332-H315-H319
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.