missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tellurium(IV) chloride, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1228422

 View more versions of this product

Catalog No. AA1228422



Specifications

Specifications

Tellurium(IV) chloride
99.9% (metals basis)
MFCD00011262
SWLJJEFSPJCUBD-UHFFFAOYSA-N
269.4
269.41
100g
10026-07-0
Cl4Te
tellurium tetrachloride, telluric chloride, tellurium chloride, tetrachlorotellurium, tellurium iv chloride, tellurium chloride tecl4 , t-4, unii-dny2r5498h, tellurium chloride, t-4, telluricchloride, tecl4
Cl[Te](Cl)(Cl)Cl
61443
Crystalline

Chemical Identifiers

10026-07-0
269.4
SWLJJEFSPJCUBD-UHFFFAOYSA-N
61443
Cl4Te
MFCD00011262
tellurium tetrachloride, telluric chloride, tellurium chloride, tetrachlorotellurium, tellurium iv chloride, tellurium chloride tecl4 , t-4, unii-dny2r5498h, tellurium chloride, t-4, telluricchloride, tecl4
Cl[Te](Cl)(Cl)Cl
Safety and Handling

Safety and Handling

EINECSNumber : 233-055-6

Documents