missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tellurium(IV) bromide, 99.9% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 07510109

 View more versions of this product

Catalog No. AA7510109



Description

Description

Used as organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

420°C (decomposition)
380°C
Crystalline
(metals basis)
MFCD00049575
14,9125
Decomposes in water forming TeO2 and HBr
Br[Te](Br)(Br)Br
447.22
447.27
Moisture sensitive
Tellurium (IV) bromide
10031-27-3
10 g
Br4Te
UN2923
tellurium tetrabromide, tellurium iv bromide, tellurium bromide tebr4 , t-4, unii-7a29efj1af, 7a29efj1af, tetrabromo-, e4-tellane, tebr4, acmc-1bozh, tellurium iv bromide, ultra dry
PTYIPBNVDTYPIO-UHFFFAOYSA-N
tetrabromo-λ⁴-tellane
82311
99.9%
4.31 g/mL at 15°C

Chemical Identifiers

10031-27-3
447.22
PTYIPBNVDTYPIO-UHFFFAOYSA-N
82311
Br[Te](Br)(Br)Br
Br4Te
MFCD00049575
tellurium tetrabromide, tellurium iv bromide, tellurium bromide tebr4 , t-4, unii-7a29efj1af, 7a29efj1af, tetrabromo-, e4-tellane, tebr4, acmc-1bozh, tellurium iv bromide, ultra dry
tetrabromo-λ⁴-tellane
Safety and Handling

Safety and Handling

GHS H Statement
H300-H314-H318
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301+H311-H314-H330-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

EINECSNumber : 233-090-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only