missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tellurium(IV) bromide, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 7510118

 View more versions of this product

Catalog No. AA7510118



Specifications

Specifications

Tellurium (IV) bromide
(metals basis)
MFCD00049575
14,9125
Decomposes in water forming TeO2 and HBr
Br[Te](Br)(Br)Br
82311
Crystalline
4.31g/mL at 15°C
Moisture Sensitive
380°C
10031-27-3
Br4Te
UN2923
tellurium tetrabromide, tellurium bromide tebr4 , t-4, tellurium iv bromide, tellurium bromide tebr4 , t-4, unii-7a29efj1af, 7a29efj1af, tetrabromo-, e4-tellane, tebr4, acmc-1bozh, tellurium iv bromide, ultra dry
PTYIPBNVDTYPIO-UHFFFAOYSA-N
447.216
447.27
99.9%
−4 Mesh
420°C (decomposition)
50g

Chemical Identifiers

10031-27-3
447.216
PTYIPBNVDTYPIO-UHFFFAOYSA-N
82311
Br4Te
MFCD00049575
tellurium tetrabromide, tellurium bromide tebr4 , t-4, tellurium iv bromide, tellurium bromide tebr4 , t-4, unii-7a29efj1af, 7a29efj1af, tetrabromo-, e4-tellane, tebr4, acmc-1bozh, tellurium iv bromide, ultra dry
Br[Te](Br)(Br)Br
Safety and Handling

Safety and Handling

GHS H Statement
H300-H314-H318
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

TSCA : TSCA

SDS Documents