Please login to your online account to display your discounted pricing

Tellurium ICP Standard, 1mL = 1mg Te (1,000ppm Te)Te in 3% HNO3, Ricca Chemical

$143.71 - $143.71

Chemical Identifiers

Chemical Name or Material Tellurium ICP Standard
CAS 7732-18-5
CAS Min % 91.0
CAS Max % 95.0
CAS 7697-37-2
CAS Min % 6.84
CAS Max % 7.12
CAS 13494-80-9
CAS Min % 0.09
CAS Max % 0.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PTE1KN100

 
ricca chemical company
RIPTE1KN100
Poly natural 100mL Each for $143.71

PTE1KN500

 
ricca chemical company
RIPTE1KN500
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 100mL
Physical Form Liquid
Color Clear colorless
Concentration or Composition (by Analyte or Components) 1mL = 1mg Te (1, 000ppm Te)Te in 3% HNO3
CAS Min % 91.0
Chemical Name or Material Tellurium ICP Standard
Packaging Poly natural
Shelf Life 18 months
CAS 7697-37-2
CAS 13494-80-9
CAS 7732-18-5
CAS Max % 95.0
CAS Max % 0.09
CAS Min % 6.84
CAS Min % 0.09
CAS Max % 7.12
Safety and Handling

18 months