Please login to your online account to display your discounted pricing

Tellurium ICP Standard, 1mL = 1mg Te (1,000ppm Te), Ricca Chemical

Chemical Identifiers

Chemical Name or Material Tellurium ICP Standard
CAS 7732-18-5
CAS Min % 92.0
CAS Max % 96.0
CAS 7647-01-0
CAS Min % 5.28
CAS Max % 5.49
CAS 13494-80-9
CAS Min % 0.09
CAS Max % 0.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PTE1KH-100

 
ricca chemical company
RIPTE1KH100
Poly natural 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PTE1KH-500

 
ricca chemical company
RIPTE1KH500
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 100mL
Concentration or Composition Notes 1,000ppm Te
Chemical Name or Material Tellurium ICP Standard
CAS 7732-18-5
CAS Max % 96.0
CAS 7647-01-0
CAS Min % 92.0
Equivalence 1 mL = 1 mg Te
Packaging Poly natural
CAS Max % 5.49
CAS 13494-80-9
CAS Min % 0.09
CAS Min % 5.28
CAS Max % 0.1