missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tellurium broken ingot, Puratronic™, 99.9999+% (metals basis)

Manufacturer:  Alfa Aesar™ 1260736

 View more versions of this product

Catalog No. AA1260736



Specifications

Specifications

Tellurium broken ingot
13494-80-9
Te
and compounds
[Te]
Odorless
6327182
Ingot
Puratronic™
500 g
(metals basis)
MFCD00134062
PORWMNRCUJJQNO-UHFFFAOYSA-N
tellurium
127.6
CHEBI:30452
≥99.9999%

Chemical Identifiers

13494-80-9
127.6
PORWMNRCUJJQNO-UHFFFAOYSA-N
6327182
tellurium
Te
MFCD00134062
and compounds
CHEBI:30452
[Te]
Safety and Handling

Safety and Handling

GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.

P264b-P270-P301+P310-P330-P501c

H301

EINECSNumber : 236-813-4

RTECSNumber : WY2625000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents