Please login to your online account to display your discounted pricing

Tellurium AA Standard, 1mL = 1mg Te (1,000ppm Te)Te in 3% HNO3, Ricca Chemical

$68.27 - $68.27

Chemical Identifiers

Chemical Name or Material Tellurium AA Standard
CAS 7732-18-5
CAS Min % 91.0
CAS Max % 95.0
CAS 7697-37-2
CAS Min % 6.84
CAS Max % 7.12
CAS 13494-80-9
CAS Min % 0.09
CAS Max % 0.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

ATE1KN100

 
ricca chemical company
RIATE1KN100
Poly natural 100mL Each for $68.27

ATE1KN500

 
ricca chemical company
RIATE1KN500
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 100mL
Color Clear colorless
Physical Form Liquid
Concentration or Composition (by Analyte or Components) 1mL = 1mg Te (1, 000ppm Te)Te in 3% HNO3
Chemical Name or Material Tellurium AA Standard
Shelf Life 24 months
CAS Max % 0.09
CAS 7732-18-5
CAS Min % 91.0
CAS Max % 95.0
CAS 7697-37-2
Packaging Poly natural
CAS Min % 6.84
CAS Max % 7.12
CAS 13494-80-9
CAS Min % 0.09
Safety and Handling

24 months