Learn More
Tellurium, 99+%, (trace metal basis), powder, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 420021000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
990.0°C | |
450.0°C | |
13494-80-9 | |
Powder | |
99+% | |
Te | |
15,298 | |
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds | |
PORWMNRCUJJQNO-UHFFFAOYSA-N | |
tellurium | |
6327182 | |
127.6 | |
Trace Metal Basis |
Tellurium | |
99+% | |
Black to Gray | |
100 g | |
Te | |
MFCD00134062 | |
15, 9263 | |
Solubility in water: insoluble. Other solubilities: soluble in h2so4, hno3 and koh, insoluble in benzene and carbon disulfide | |
[Te] | |
127.60 | |
CHEBI:30452 | |
≥99% | |
Glass bottle |
Chemical Identifiers
13494-80-9 | |
127.60 | |
PORWMNRCUJJQNO-UHFFFAOYSA-N | |
6327182 | |
tellurium |
Te | |
MFCD00134062 | |
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds | |
CHEBI:30452 | |
[Te] |
Safety and Handling
GHS H Statement
Harmful if inhaled.
May cause an allergic skin reaction.
May cause long lasting harmful effects to aquatic life.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position
GHS Signal Word: Warning
EINECSNumber : 236-813-4
RTECSNumber : WY2625000
TSCA : TSCA
RUO – Research Use Only