Please login to your online account to display your discounted pricing

Tellurium, 99.8%, (trace metal basis), powder, -200 mesh, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC315995000

 View more versions of this product

Catalog No. AC315995000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Tellurium
CAS 13494-80-9
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Rinse mouth.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Purity Grade Trace Metal Basis
Physical Form Powder
Assay Percent Range 99.8%
Molecular Formula Te
Formula Weight 127.6
MDL Number MFCD00134062
Fieser 15,298
Merck Index 15, 9263
Packaging Glass bottle
Total Trace Metal Impurities 0.2% max.
Color Gray to black
Quantity 500g

Specifications

Chemical Name or Material Tellurium
CAS 13494-80-9
Purity Grade Trace Metal Basis
Physical Form Powder
Assay Percent Range 99.8%
Molecular Formula Te
Formula Weight 127.6
MDL Number MFCD00134062
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Rinse mouth.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.

SDS & Safety Documents