Please login to your online account to display your discounted pricing

Tellurium, 99.8%, (trace metal basis), powder, -200 mesh, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  315991000

 View more versions of this product

Catalog No. AC315991000

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Trace Metal Basis
Formula Weight 127.6
Physical Form Powder
Quantity 100g
Color Gray to black
Fieser 15,298
Merck Index 15, 9263
CAS 13494-80-9
Assay Percent Range 99.8%
Molecular Formula Te
Chemical Name or Material Tellurium
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
MDL Number MFCD00134062
Packaging Glass bottle
Total Trace Metal Impurities 0.2% max.
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Rinse mouth.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Tellurium
CAS 13494-80-9
Purity Grade Trace Metal Basis
Physical Form Powder
Assay Percent Range 99.8%
Molecular Formula Te
Formula Weight 127.6
MDL Number MFCD00134062
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Rinse mouth.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.

SDS & Safety Documents