missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Telluric acid dihydrate, >99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1419718

 View more versions of this product

Catalog No. AA1419718



Specifications

Specifications

13451-14-4
H6O6Te
telluric acid dihydrate, telluricacidwhitepowder, telluric acid-te
229.64
Crystalline
50g
>99%
MFCD00150387
VCYUGZHEVVXOPD-UHFFFAOYSA-N
229.64 (193.61 Anhydrous)
White

Chemical Identifiers

13451-14-4
229.64
VCYUGZHEVVXOPD-UHFFFAOYSA-N
H6O6Te
MFCD00150387
telluric acid dihydrate, telluricacidwhitepowder, telluric acid-te
SDS Documents