TEK (TIE2) [Y1108F] Recombinant Human Protein, Invitrogen™ DFS Item
Human TEK (TIE2) Y1108F, C-terminal fragment, amino acids Q771-A1124 (as in NCBI/Protein entry NP_000450.2) with a Y1108F mutation, N-terminal GST-HIS6 fusion protein with a 3C cleavage site, expressed in Sf9 insect cells.
Manufacturer:  Life Technologies LSPV6229
 View more versions of this product
Catalog No. PV6229
Description
Description
  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications
Specifications
Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
69.0kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Kinase Assay
Human
GST Tag, His Tag
50mM HEPES pH 7.5, 100mM NaCl, 5mM DTT, 15mM reduced glutathione, 20% glycerol
10μg
TEK, TIE-2
aa 771-1124
Yes
Documents