Invitrogen™ TEK (TIE2) Recombinant Human Protein DFS Item
The TEK receptor tyrosine kinase is expressed almost exclusively in endothelial cells.
Manufacturer:  Invitrogen™ LSPV4199
 View more versions of this product
Catalog No. PV4199
Description
Description
TEK signaling appears to be critical for endothelial cell-smooth muscle cell communication in venous morphogenesis, and defects in TEK are associated with inherited venous malformations.
Specifications
Specifications
NP_000450
Frozen
61.5kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
Kinase Assay
TIE Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
TEK (Tie2)
Baculovirus
Human
GST Tag
Documents