missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ TEK (TIE2) Recombinant Human Protein DFS Item

The TEK receptor tyrosine kinase is expressed almost exclusively in endothelial cells.

Manufacturer:  Invitrogen™ LSPV3628

 View more versions of this product

Catalog No. PV3628



Description

Description

TEK signaling appears to be critical for endothelial cell-smooth muscle cell communication in venous morphogenesis, and defects in TEK are associated with inherited venous malformations.

Specifications

Specifications

NP_000450
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
61.5kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Recombinant, Cytoplasmic
TEK (Tie2)
Kinase Assay
Baculovirus
aa 817-1101
Yes
Cell Viability, Proliferation & Function|Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Cell Analysis|Cell Signaling|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
7010, 7124
10μg
RUO - research use only
Receptor Tyrosine Kinases
TEK tyrosine kinase, endothelial
Kinases & Inhibitors
TIE Kinase Family
Human
GST Tag
Documents