Please login to your online account to display your discounted pricing

Tocris™ Teijin compound 1

Potent CCR2b antagonist

$252.39 - $1,050.42

Specifications

Product Type Teijin compound 1
Inhibitors Teijin compound 1
Molecular Weight 476.32
Purity >98
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

366410R

 
tocris™
366410
10mg Each for $252.39

366450R

 
tocris™
366450
50mg Each for $1,050.42
Description & Specifications

Specifications

Product Type Teijin compound 1
Inhibitors Teijin compound 1
Molecular Weight 476.32
Purity >98

Potent chemokine CCR2b receptor antagonist (IC50 = 180nM). Potently inhibits cell chemotaxis induced by MCP-1 (EC50 = 24nM).

  • Chemical Name: N-[2-[[(3R)-1-[(4-chlorophenyl)methyl]-3-pyrrolidinyl]amino]-2-oxoethyl]-3-(trifluoromethyl)benzamide hydrochloride
  • Formula: C21H21ClF3N3O2.HCl
  • CAS Number: 1313730-14-1
  • Solubility: Soluble to 10mM in water and to 100mM in DMSO
  • Purity: >98