missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tecniplast™ Stainless Steel Wire Cage Lids and Feeders
Secure and easty to handle
Supplier: Tecniplast™ 1354G117
Description
- U-shaped hopper allows easy access to food and won't entrap food or contaminants
- Cross-wire members prevent bending
- 0.07cm wire spacing
- Folding hopper divider allows lids to be stacked for storage
- Central divider prevents animal from crawling into feeder
- Can be used with or without a raised wire floor
- Autoclavable
Specifications
Stainless Steel | |
Rats and Hamsters | |
0.07 cm |
Wire Cage Lid with Feeder | |
Cage size G |