Please login to your online account to display your discounted pricing

Techne™ Filter Cartridges for PrimeQ Real-time PCR System


Incorporates both excitation and emission filters together with dichroic mirror for fluorophore excitation and emission detection

Manufacturer: techne™  FC05

 View more versions of this product

Catalog No. 13-245-152

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Dye Type Cy5, Quasar 670
Excitation Value 640λ
Kit Type Filter Cartridge
Wavelength 685λ