Please login to your online account to display your discounted pricing

Techne™ Filter Cartridges for PrimeQ Real-time PCR System


Incorporates both excitation and emission filters together with dichroic mirror for fluorophore excitation and emission detection

Manufacturer: techne™  FC04

 View more versions of this product

Catalog No. 13-245-151

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Dye Type ROX, TEXAS Red, Cy 3.5
Excitation Value 580λ
Kit Type Filter Cartridge
Wavelength 615λ