Please login to your online account to display your discounted pricing

Techne™ Filter Cartridges for PrimeQ Real-time PCR System


Incorporates both excitation and emission filters together with dichroic mirror for fluorophore excitation and emission detection

Manufacturer: techne™  FC03

 View more versions of this product

Catalog No. 13-245-150

  • / Each
 

Description & Specifications

Specifications

Kit Type Filter Cartridge
Dye Type HEX, TET, JOE, VIC, Yakima Yellow
Excitation Value 530λ
Wavelength 560λ