Please login to your online account to display your discounted pricing

Techne™ Filter Cartridges for PrimeQ Real-time PCR System


Incorporates both excitation and emission filters together with dichroic mirror for fluorophore excitation and emission detection

Manufacturer: techne™  FC02

 View more versions of this product

Catalog No. 13-245-149

  • / Each
 

Description & Specifications

Specifications

Kit Type Filter Cartridge
Wavelength 520λ
Dye Type FAM, green intercalating dyes, Fluorescein, EvaGreen
Excitation Value 485λ