Please login to your online account to display your discounted pricing
Techne™ 5Prime Thermal Cyclers
Delivers both high performance and high throughput to provide maximum flexibility when processing large number of samples in parallel and ability to upgrade base model to gradient capability
Manufacturer: techne™  5PRIME02
 View more versions of this product
Catalog No. 13-245-132
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Capacity 96 x 0.2mL
Electrical Requirements 100-230V 50-60Hz, 450w
Product Type Prime thermal cycler