missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tech Glass Squeeze-O-Matic Burets with Bottles and PTFE Plugs

Automatic self-zeroring burets come with base plates for stability

$403.07 - $403.07

Specifications

Color White
For Use With (Application) Used with acidic solutions
Includes LDPE bottle, glass plug, base plate for added stability
Material LDPE
Stopcock Material Glass
View More Specs

Includes: Low-density polyethylene bottle, glass plug, base plate for added stability

Products 1
Catalog Number Mfr. No. Capacity (Metric) For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) For Use With (Equipment) Price Quantity  
10-436-23 View Documents Fisher Scientific
TG184352520*
25 mL 1000mL LDPE bottle and glass plug Each for $403.07
 
Description

Description

  • Automatic self-zeroing
  • Can be used with alkaline solutions
  • Color: White

Specifications

Specifications

White
LDPE bottle, glass plug, base plate for added stability
Glass
Used with acidic solutions
LDPE