Please login to your online account to display your discounted pricing

Tech Glass Squeeze-O-Matic Burets with Bottles and PTFE Plugs


Automatic self-zeroring burets come with base plates for stability

Manufacturer: fisher scientific  TG184352516

 View more versions of this product

Catalog No. 10-436-22

  • / Each
 

Description & Specifications

Specifications

Capacity (Metric) 25mL
For Use With (Application) Used with acidic solutions
For Use With (Equipment) 500mL LDPE bottle and glass plug
Includes LDPE bottle, glass plug, base plate for added stability
Material LDPE
Stopcock Material Glass
Color White