Please login to your online account to display your discounted pricing

Tech Glass Squeeze-O-Matic Burets with Bottles and Glass Plugs

Automatic self-zeroring burets come with base plates for stability

$331.03 - $331.03

Specifications

Color White
For Use With (Application) Used with acidic solutions
Includes LDPE bottle, glass plug, base plate for added stability
Material LDPE
Stopcock Material Glass
View More Specs
Includes:  Low-density polyethylene bottle, glass plug, base plate for added stabliity
Products
Catalog Number Mfr. No. For Use With (Equipment) Capacity (Metric) Price Quantity    

10-436-31

 
fisher scientific
TG184301019
1000mL LDPE bottle and glass plug 10mL Each for $331.03
Description & Specifications

Specifications

Color White
For Use With (Application) Used with acidic solutions
Includes LDPE bottle, glass plug, base plate for added stability
Material LDPE
Stopcock Material Glass

  • Automatic self-zeroing
  • Can be used with Acidic solutions
  • Color: White