Please login to your online account to display your discounted pricing
Tech Glass Replacement Squeeze-O-Matic Burets with PTFE Plugs
Replacement buret with PTFE plug
$235.77 - $249.13
Specifications
For Use With (Application) Used with acidic solutions
View More Specs
Products
Catalog Number Mfr. No. For Use With (Equipment) Capacity (Metric) Price Quantity    
10-436-27
fisher scientific
TG184152516
500mL LDPE bottle 25mL Each for $249.13
10-436-25
fisher scientific
TG184151016
500mL LDPE bottle 10mL Each for $235.77
Description & Specifications
Specifications
For Use With (Application) Used with acidic solutions
  • Automatic self-zeroing
  • Used with low density polyethylene bottle
  • Used with alkaline solutions