Please login to your online account to display your discounted pricing
Tech Glass Replacement Squeeze-O-Matic Burets with PTFE Plugs
Replacement buret with PTFE plug
Manufacturer: fisher scientific  TG184151016
 View all options for this product
Catalog No. 10-436-25
Description & Specifications
Capacity (Metric) 10mL
For Use With (Application) Used with acidic solutions
For Use With (Equipment) 500mL LDPE bottle
  • Automatic self-zeroing
  • Used with low density polyethylene bottle
  • Used with alkaline solutions