Please login to your online account to display your discounted pricing

Tebbe reagent, 0.5M solution in toluene, ACROS Organics™


Quantity 25mL
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  AC368970250

 View more versions of this product

Catalog No. AC368970250

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Solution
Formula Weight 284.62
Quantity 25mL
Color Dark red to purple
Packaging Glass Bottle
Concentration 0.40 to 0.60M
Density 0.9270g/mL
Fieser 08,83; 10,87; 11,52; 12,54; 13,71; 15,79; 16,70; 17,69
Chemical Name or Material Tebbe reagent
MDL Number MFCD00151575
Health Hazard 1 Danger
CAS 108-88-3
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Suspected of damaging fertility or the unborn child.
Highly flamma
Name Note 0.5M solution in toluene
CAS 67719-69-1
Synonym Bis(cyclopentadienyl-μ-chloro(dimethylaluminum-μ-methylenetitanium
Molecular Formula C13H18AlClTi

Specifications

Chemical Name or Material Tebbe reagent
Name Note 0.5M solution in toluene
CAS 67719-69-1
CAS 108-88-3
Molecular Formula C13H18AlClTi
Formula Weight 284.62
MDL Number MFCD00151575
Synonym Bis(cyclopentadienyl-μ-chloro(dimethylaluminum-μ-methylenetitanium
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Suspected of damaging fertility or the unborn child.
Highly flamma

SDS & Safety Documents