Please login to your online account to display your discounted pricing

Teacher's Shears


Multi-purpose and very sharp.

Manufacturer: fisher science education  101C7

 View more versions of this product

Catalog No. S173172

  • / Each
 

Description & Specifications

Specifications

Length (English) 7 in.
For Use With (Application) Office
Blade Style Straight
Length (Metric) 20.3cm