missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ TEA, 0.2M buffer soln., pH 8.5

Manufacturer:  Alfa Aesar™ J61668AK

 View more versions of this product

Catalog No. AAJ61668AK



Specifications

Specifications

TEA buffer soln.
C6H15NO3
0.2M buffer soln.
GSEJCLTVZPLZKY-UHFFFAOYSA-N
2-[bis(2-hydroxyethyl)amino]ethanol
7618
Liquid
8.5
102-71-6
MFCD00002855
Triethanolamine buffer solution
C(CO)N(CCO)CCO
149.19
CHEBI:28621
Odorless
250mL

Chemical Identifiers

102-71-6
149.19
GSEJCLTVZPLZKY-UHFFFAOYSA-N
7618
2-[bis(2-hydroxyethyl)amino]ethanol
C6H15NO3
MFCD00002855
Triethanolamine buffer solution
CHEBI:28621
C(CO)N(CCO)CCO
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents