missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ TEA, 0.2M buffer soln., pH 7.5

$84.96 - $84.96

Chemical Identifiers

CAS 102-71-6
Molecular Formula C6H15NO3
Molecular Weight (g/mol) 149.19
MDL Number MFCD00002855
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
Synonym Triethanolamine buffer solution
PubChem CID 7618
ChEBI CHEBI:28621
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethanol
SMILES C(CO)N(CCO)CCO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61319AK View Documents Alfa Aesar™
J61319AK
250mL Each for $84.96
 
Specifications

Chemical Identifiers

102-71-6
149.19
GSEJCLTVZPLZKY-UHFFFAOYSA-N
7618
2-[bis(2-hydroxyethyl)amino]ethanol
C6H15NO3
MFCD00002855
Triethanolamine buffer solution
CHEBI:28621
C(CO)N(CCO)CCO

Specifications

102-71-6
MFCD00002855
Triethanolamine buffer solution
C(CO)N(CCO)CCO
149.19
CHEBI:28621
Odorless
250mL
C6H15NO3
0.2M buffer soln.
GSEJCLTVZPLZKY-UHFFFAOYSA-N
2-[bis(2-hydroxyethyl)amino]ethanol
7618
Liquid
7.5
TEA buffer soln.
Safety and Handling

Safety and Handling

TSCA : TSCA