Please login to your online account to display your discounted pricing

TE Buffer, 100X, Molecular Biology Grade, Calbiochem™, EMD Millipore

A 100X concentrate that, when diluted to 1X, contains 10mM Tris and 1mM EDTA

$196.01 - $196.01

Chemical Identifiers

Analyte or Component Names Tris, EDTA
CAS 1185-53-1
Chemical Name or Material TE Buffer
Concentration or Composition (by Analyte or Components) 1X, contains 10mM Tris, 1mM EDTA
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5747931L

 
emd millipore corporation
5747931L
Each for $196.01
Description & Specifications

Specifications

Quantity 1L
Chemical Name or Material TE Buffer
Analyte or Component Names Tris, EDTA
CAS 1185-53-1
DNase None Detected
Synonym Tris-EDTA Buffer
Concentration or Composition (by Analyte or Components) 1X, contains 10mM Tris, 1mM EDTA
CAS 60-00-4
Protease None Detected
Purity Molecular Biology Grade
Recommended Storage 15°C to 30°C (Do not freeze); Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.
RNase None Detected
Safety and Handling

15°C to 30°C (Do not freeze); Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.