missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ TdT/Terminal Deoxyribonucleotidyl Transferase Ab-1, Mouse Monoclonal Antibody

TdT/Terminal Deoxyribonucleotidyl Transferase Ab-1, Mouse MAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.

$298.31 - $1,370.08

Specifications

Antigen TdT/Terminal Deoxyribonucleotidyl Transferase Ab-1
Clone SEN28
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MS1105R7 Thermo Scientific™ Lab Vision™
MS1105R7
7mL Each for $793.27
MS1105S Thermo Scientific™ Lab Vision™
MS1105S
1mL Each for $1,370.08
MS1105S0 Thermo Scientific™ Lab Vision™
MS1105S0
100μL Each for $298.31
MS1105S1 Thermo Scientific™ Lab Vision™
MS1105S1
500μL Each for $919.18
Specifications

Description

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Thermo Scientific™ TdT/Terminal Deoxyribonucleotidyl Transferase Ab-1, Mouse Monoclonal Antibody.

Terminal Deoxynucleotidyl Transferase (TdT) is a DNA polymerase located in the cell nucleus which catalyses the polymerization of deoxynucleotides at the 3' hydroxyl ends of oligo or polydeoxynucleotide initiators and functions without a template. TdT is considered to be a highly specific marker for the diagnosis and classification of acute lymphoblastic lymphoma/leuksemias. The determination of TdT expression is most valuable when it is different to differentiate histologically between lymphoblastic lymphoma and Burkitt's lymphoma.

Host Species: Mouse

Clone: SEN28

Isotype: IgG2a

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: N-terminal

Immunogen: Recombinant protein corresponding to the amino terminal region of the TdT molecule

Molecular Weight: 58kDa

Positive Control: Thymus

Cellular Localization: Nuclear Ab-1 will be of use in the differentiation of lymphoblastic lymphoma from other lymphomas.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)


Description & Specifications

Specifications

TdT/Terminal Deoxyribonucleotidyl Transferase Ab-1
Immunohistochemistry (Paraffin)
Unlabeled
IVD
Primary
SEN28
Monoclonal
Murine
DNA Repair
Recombinant protein corresponding to the amino terminal region of the TdT molecule