missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TdT Mouse, Clone: 7UNA8U6, eBioscience™

Mouse Monoclonal Antibody

$146.45 - $361.05

Specifications

Antigen TdT
Clone 7UNA8U6
Host Species Murine
Gene Alias Terminal-deoxynucleotidyl transferase
Species Reactivity Human, Mouse, Rat
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122805 Life Technologies
LS14973980
25μg Each for $146.45
501122806 Life Technologies
LS14973982
100μg Each for $361.05
Specifications

Description

The monoclonal antibody 7UNA8U6 reacts with human, mouse, and rat TdT. TdT, or terminal-deoxynucleotidyl transferase, is a 60kDa polymerase responsible for the template-independent addition of N-nucleotides at gene segment junctions of developing lymphocytes. Expression is found in the nucleus of immature lymphocytes but not in mature lymphocytes or non-lymphoid cells. TdT plays a critical role during TCR and Ig gene rearrangement and functions to increase the diversity of the lymphocyte repertoire through the addition of N-nucleotides onto the ends of newly formed P-nucleotides. After positive selection, expression of TdT becomes decreased or absent. Expression of TdT has been shown to be present in lymphoma or lymphoblastic leukemia cells.
Description & Specifications

Specifications

TdT
Murine
Human, Mouse, Rat
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
7UNA8U6
Terminal-deoxynucleotidyl transferase
Immunohistochemistry,Immunohistochemistry (Formalin/Paraffin),Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL