missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TdT Mouse anti-Mouse, PE, Clone: 19-3, eBioscience™

Mouse Monoclonal Antibody

$170.28 - $394.68

Specifications

Antigen TdT
Clone 19-3
Host Species Murine
Gene Alias terminal deoxynucleotidyl transferase
Applications Flow Cytometry (Intracellular Staining)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010902 Life Technologies
LS12584680
25μg Each for $170.28
5010903 Life Technologies
LS12584682
100μg Each for $394.68
Specifications

Description

This 19-3 monoclonal antibody recognizes human and mouse terminal-deoxynucleotidyl transferase (TdT), a 60kDa polymerase responsible for the template-independent addition of N-nucleotides at gene segment junctions of developing lymphocytes. Expression is found in the nucleus of immature lymphocytes but not in mature lymphocytes or non-lymphoid cells. Tdt plays a critical role during TCR and Ig gene rearrangement. Regulation of expression in the mouse thymus correlates with T cell selection; decreased or absent after positive selection. Additionally Tdt has been shown to be present in lymphoma or lymphoblastic leukemia cells.
Description & Specifications

Specifications

TdT
Murine
Flow Cytometry (Intracellular Staining)
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
19-3
terminal deoxynucleotidyl transferase
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer