missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TCS PrP Inhibitor 13, Tocris

Antiprion agent; potently inhibits PrP-res accumulation

$187.24 - $789.24

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C15H11N3O3
Formula Weight 281.27
Synonym 2, 4-Dihydro-5-(4-nitrophenyl)-2-phenyl-3H-pyrazol-3-one
Chemical Name or Material TCS PrP Inhibitor 13
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
323810 R & D Systems
RDS323810
10mg Each for $187.24
323850 R & D Systems
RDS323850
50mg Each for $789.24
Specifications Description & Specifications

Chemical Identifiers

>98%
281.27
TCS PrP Inhibitor 13
C15H11N3O3
2, 4-Dihydro-5-(4-nitrophenyl)-2-phenyl-3H-pyrazol-3-one

Specifications

10mg
>98%
281.27
TCS PrP Inhibitor 13
34320-83-7
C15H11N3O3
2, 4-Dihydro-5-(4-nitrophenyl)-2-phenyl-3H-pyrazol-3-one

Safety and Handling

Recommended Storage : Store at +4°C