missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tocris™ TCS PIM-1 4a

Selective, ATP-competitive Pim kinase inhibitor

$125.42 - $530.81

Specifications

Product Type TCS PIM-1 4a
Inhibitors TCS PIM-1 4a
Molecular Weight 273.23
Purity >98
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
371410R Tocris™
RDS371410
10mg Each for $125.42
371450R Tocris™
RDS371450
50mg Each for $530.81
Specifications

Description

Selective, ATP-competitive Pim kinase inhibitor (IC50 values are 24 and 100nM for Pim-1 and Pim-2 respectively). Displays selectivity over a panel of ∽50 other kinases tested. Exhibits cytotoxicity in PC3 prostate carcinoma cells in vitro (IC50 = 17μM).
  • Chemical Name: (5Z)-5-[[3-(Trifluoromethyl)phenyl]methylene]-2,4-thiazolidinedione
  • Formula: C11H6F3NO2S
  • CAS Number: 438190-29-5
  • Solubility: Soluble to 100mM in DMSO and to 100mM in ethanol
  • Purity: >98
Description & Specifications

Specifications

TCS PIM-1 4a
TCS PIM-1 4a
273.23
>98