Please login to your online account to display your discounted pricing

TCS PIM-1 1, Tocris

Selective, ATP-competitive Pim-1 kinase inhibitor

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material TCS PIM-1 1
Assay Percent Range >98%
Molecular Formula C18H11BrN2O2
Formula Weight 367.2
Synonym 3-Cyano-4-phenyl-6-(3-bromo-6-hydroxy)phenyl-2(1H)-pyridone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

297910

 
r & d systems
RDS297910
10mg Each for $201.36

297950

 
r & d systems
RDS297950
50mg Each for $844.73
Description & Specifications

Specifications

Chemical Name or Material TCS PIM-1 1
Quantity 10mg
CAS 491871-58-0
Recommended Storage Store at +4°C
Assay Percent Range >98%
Molecular Formula C18H11BrN2O2
Formula Weight 367.2
Synonym 3-Cyano-4-phenyl-6-(3-bromo-6-hydroxy)phenyl-2(1H)-pyridone
Safety and Handling

Store at +4°C