Please login to your online account to display your discounted pricing

TCS OX2 29, Tocris

Potent and selective OX2 antagonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material TCS OX2 29
Assay Percent Range >98%
Molecular Formula C23H31N3O3·HCl
Formula Weight 433.97
Synonym (2S)-1-(3,4-Dihydro-6,7-dimethoxy-2(1H)-isoquinolinyl)-3,3-dimethyl-2-[(4-pyridinylmethyl)amino]-1-butanone hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

337150

 
r & d systems
RDS337150
50mg Each for $1,050.42

337110

 
r & d systems
RDS337110
10mg Each for $252.39
Description & Specifications

Specifications

Formula Weight 433.97
Quantity 50mg
Chemical Name or Material TCS OX2 29
Assay Percent Range >98%
Molecular Formula C23H31N3O3·HCl
CAS 372523-75-6
Synonym (2S)-1-(3,4-Dihydro-6,7-dimethoxy-2(1H)-isoquinolinyl)-3,3-dimethyl-2-[(4-pyridinylmethyl)amino]-1-butanone hydrochloride
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C