Please login to your online account to display your discounted pricing

TCS 5861528, Tocris™

TRPA1 blocker

$195.49 - $816.90

Chemical Identifiers

Chemical Name or Material TCS 5861528
Assay Percent Range >98%
Molecular Formula C19H23N5O3
Formula Weight 369.42
Synonym 2-(1,3-Dimethyl-2,6-dioxo-1,2,3,6-tetrahydro-7H-purin-7-yl)-N-[4-(1-methylpropyl)phenyl]acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

393850

 
r & d systems
393850
50mg Each for $816.90

393810

 
r & d systems
393810
10mg Each for $195.49
Description & Specifications

Specifications

Formula Weight 369.42
Quantity 50mg
Chemical Name or Material TCS 5861528
Recommended Storage Store at Room Temperature
Synonym 2-(1,3-Dimethyl-2,6-dioxo-1,2,3,6-tetrahydro-7H-purin-7-yl)-N-[4-(1-methylpropyl)phenyl]acetamide
Assay Percent Range >98%
CAS 332117-28-9
Molecular Formula C19H23N5O3
Safety and Handling

Store at Room Temperature