Please login to your online account to display your discounted pricing

TCS 46b, Tocris

Orally active, subtype-selective NR1A/NR2B antagonist

$212.74 - $886.68

Chemical Identifiers

Chemical Name or Material TCS 46b
Assay Percent Range >98%
Molecular Formula C22H23N3O
Formula Weight 345.44
Synonym 1, 3-Dihydro-5-[3-[4-(phenylmethyl)-1-2H-benzimidazol-2-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

278250

 
r & d systems
RDS278250
50mg Each for $886.68

278210

 
r & d systems
RDS278210
10mg Each for $212.74
Description & Specifications

Specifications

Formula Weight 345.44
Quantity 50mg
Chemical Name or Material TCS 46b
Assay Percent Range >98%
Molecular Formula C22H23N3O
CAS 302799-86-6
Synonym 1, 3-Dihydro-5-[3-[4-(phenylmethyl)-1-2H-benzimidazol-2-one
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature