Please login to your online account to display your discounted pricing

TCS 21311, Tocris™

Potent JAK3 inhibitor; Also inhibits GSK-3β and PKC

$297.57 - $1,231.36

Chemical Identifiers

Chemical Name or Material TCS 21311
Assay Percent Range >98%
Molecular Formula C27H25F3N4O4
Formula Weight 526.51
Synonym 3-[5-[4-(2-Hydroxy-2-methyl-1-oxopropyl)-1-piperazinyl]-2-(trifluoromethyl)phenyl]-4-(1H-indol-3-yl)-1H-pyrrole-2, 5-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

422110

 
r & d systems
422110
10mg Each for $297.57

422150

 
r & d systems
422150
50mg Each for $1,231.36
Description & Specifications

Specifications

Formula Weight 526.51
Quantity 10mg
Molecular Formula C27H25F3N4O4
Recommended Storage Desiccate at Room Temperature
Chemical Name or Material TCS 21311
Assay Percent Range >98%
Synonym 3-[5-[4-(2-Hydroxy-2-methyl-1-oxopropyl)-1-piperazinyl]-2-(trifluoromethyl)phenyl]-4-(1H-indol-3-yl)-1H-pyrrole-2, 5-dione
CAS 1260181-14-3
Safety and Handling

Desiccate at Room Temperature