Please login to your online account to display your discounted pricing

TCS 1205, Tocris™

Subtype-selective GABAA receptor agonist

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material TCS 1205
Assay Percent Range >99%
Molecular Formula C18H15N3O4
Formula Weight 337.33
Synonym 5-Nitro-α-oxo-N-(1R)-phenylethyl]-1H-indole-3-acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

394110

 
r & d systems
394110
10mg Each for $205.70

394150

 
r & d systems
394150
50mg Each for $862.95
Description & Specifications

Specifications

Formula Weight 337.33
Quantity 10mg
Chemical Name or Material TCS 1205
Assay Percent Range >99%
Molecular Formula C18H15N3O4
CAS 355022-97-8
Synonym 5-Nitro-α-oxo-N-(1R)-phenylethyl]-1H-indole-3-acetamide
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature