Please login to your online account to display your discounted pricing

TCS 1105, Tocris™

GABAA α2 benzodiazepine receptor agonist

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material TCS 1105
Assay Percent Range >99%
Molecular Formula C17H13FN2O2
Formula Weight 296.3
Synonym N-[(-4-Fluorophenyl)methyl]-α-oxo-1H-indole-3-acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

394250

 
r & d systems
394250
50mg Each for $862.95

394210

 
r & d systems
394210
10mg Each for $205.70
Description & Specifications

Specifications

Formula Weight 296.3
Quantity 50mg
Chemical Name or Material TCS 1105
CAS 185391-33-7
Assay Percent Range >99%
Molecular Formula C17H13FN2O2
Synonym N-[(-4-Fluorophenyl)methyl]-α-oxo-1H-indole-3-acetamide
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature