Please login to your online account to display your discounted pricing

TCS 1102, Tocris™

Potent, dual orexin receptor antagonist

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material TCS 1102
Assay Percent Range >98%
Molecular Formula C27H26N4O2S
Formula Weight 470.59
Synonym N-[1, 1'-Biphenyl]-2-yl-1-[2-[(1-methyl-1H-benzimidazol-2-yl)thio]acetyl-2-pyrrolidinedicarboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

381810

 
r & d systems
381810
10mg Each for $201.36

381850

 
r & d systems
381850
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 470.59
Quantity 10mg
CAS 916141-36-1
Assay Percent Range >98%
Molecular Formula C27H26N4O2S
Synonym N-[1, 1'-Biphenyl]-2-yl-1-[2-[(1-methyl-1H-benzimidazol-2-yl)thio]acetyl-2-pyrrolidinedicarboxamide
Chemical Name or Material TCS 1102
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature