Please login to your online account to display your discounted pricing

TCR beta Armenian Hamster anti-Mouse, eFluor 450, Clone: H57-597, eBioscience™

Armenian Hamster Monoclonal Antibody

$145.00 - $324.80

Specifications

Antigen TCR beta
Clone H57-597
Host Species Armenian Hamster
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50246027

 
life technologies
LS48596180
25μg Each for $145.00

50246028

 
life technologies
LS48596182
100μg Each for $324.80
Description & Specifications

Specifications

Antigen TCR beta
Clone H57-597
Host Species Armenian Hamster
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The H57-597 monoclonal antibody reacts with the beta chain of mouse TCR. TCR beta is expressed by thymocytes in a developmentally regulated manner and a majority of peripheral T cells. Crosslinking of the TCR complex with H57-597 induces activation and proliferation of T cells or apoptosis based on assay conditions. H57-597 is used as a phenotypic marker for TCR beta expressing T cells.